1-Trifluoromethoxymethyl-cyclopropanecarboxylic acid 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC520780-1G
PC520780-250MG
PC520780-500MG
APOSPC520780-1GEA
11860
DKK
APOSPC520780-1G
APOSPC520780-250MG
APOSPC520780-500MG
1-Trifluoromethoxymethyl-cyclopropanecarboxylic acid 97%
1-Trifluoromethoxymethyl-cyclopropanecarboxylic acid
Formel:
C₆H₇F₃O₃ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD23382132 CAS-nummer: 1408279-84-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...