1-Bromo-4-[chloro(difluoro)methoxy]benzene 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC520765-1G PC520765-500MG PC520765-5G PC520765-10G
APOSPC520765-1GEA 1830 DKK
APOSPC520765-1G APOSPC520765-500MG APOSPC520765-5G APOSPC520765-10G
1-Bromo-4-[chloro(difluoro)methoxy]benzene 95%
1-Bromo-4-[chloro(difluoro)methoxy]benzene
Formel: C₇H₄BrClF₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22580792
CAS-nummer: 112556-13-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR