5-(Difluoromethyl)pyrazine-2-carboxylic acid 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC520751-1G PC520751-5G
APOSPC520751-1GEA 806 DKK
APOSPC520751-1G APOSPC520751-5G
5-(Difluoromethyl)pyrazine-2-carboxylic acid 95%
5-(Difluoromethyl)pyrazine-2-carboxylic acid
Formel: C₆H₄F₂N₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20640530
CAS-nummer: 1174321-06-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR