2-Amino-4-bromo-5-fluorobenzoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 4-Bromo-5-fluoroanthranilic acid

PC52072-1G PC52072-250MG PC52072-5G
APOSPC52072-1GEA 600 DKK
APOSPC52072-1G APOSPC52072-250MG APOSPC52072-5G
2-Amino-4-bromo-5-fluorobenzoic acid
2-Amino-4-bromo-5-fluorobenzoic acid
Formel: C₇H₅BrFNO₂
Molekylvægt: 234.023 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08702774
CAS-nummer: 1374208-42-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR