5-Fluoro-3-methyl-2-nitroaniline 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC51367-1G
PC51367-250MG
APOSPC51367-1GEA
6490
DKK
APOSPC51367-1G
APOSPC51367-250MG
5-Fluoro-3-methyl-2-nitroaniline 95%
5-Fluoro-3-methyl-2-nitroaniline
Formel:
C₇H₇FN₂O₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD27950992 CAS-nummer: 468741-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...