Methyl pentafluoropropionylacetate 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC5133-1G PC5133-5G PC5133-10G
APOSPC5133-1GEA 350 DKK
APOSPC5133-1G APOSPC5133-5G APOSPC5133-10G
Methyl pentafluoropropionylacetate 95%
Methyl pentafluoropropionylacetate
Formel: C₆H₅F₅O₃
Kogepunkt: 124…126 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041000
CAS-nummer: 104857-88-7
UN nummer: 1993
ADR: 3,III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR