3,4,5-Trifluoropyridine 97%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC51194-1G
PC51194-5G
APOSPC51194-1GEA
1910
DKK
APOSPC51194-1G
APOSPC51194-5G
3,4,5-Trifluoropyridine 97%
3,4,5-Trifluoropyridine
Formel:
C₅H₂F₃N Kogepunkt: 85…87 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09998883 CAS-nummer: 67815-54-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...