4-Amino-3-fluorobenzeneboronic acid, N-CBZ protected 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-(Benzyloxycarbonylamino)-3-fluorobenzeneboronic acid

PC5108-1G PC5108-5G
APOSPC5108-1GEA 1040 DKK
APOSPC5108-1G APOSPC5108-5G
4-Amino-3-fluorobenzeneboronic acid, N-CBZ protected 98%
4-Amino-3-fluorobenzeneboronic acid, N-CBZ protected
Formel: C₁₄H₁₃BFNO₄
Smeltepunkt: 197…199 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08235098
CAS-nummer: 874290-60-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR