6-Bromo-3-chloro-5-fluoro-1-methyl-1H-indazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC51024-1G
PC51024-250MG
PC51024-500MG
APOSPC51024-1GEA
2300
DKK
APOSPC51024-1G
APOSPC51024-250MG
APOSPC51024-500MG
6-Bromo-3-chloro-5-fluoro-1-methyl-1H-indazole
6-Bromo-3-chloro-5-fluoro-1-methyl-1H-indazole
Formel:
C₈H₅BrClFN₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD18384876 CAS-nummer: 1286734-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...