N-[[4-(4-Fluorobenzyl)-2-morpholinyl]methyl]acetamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC510236-100G PC510236-25G
APOSPC510236-100GEA 872 DKK
APOSPC510236-100G APOSPC510236-25G
N-[[4-(4-Fluorobenzyl)-2-morpholinyl]methyl]acetamide
N-[[4-(4-Fluorobenzyl)-2-morpholinyl]methyl]acetamide
Formel: C₁₄H₁₉FN₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16619692
CAS-nummer: 112913-94-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR