2-Ethyl-4-fluoro-1-nitrobenzene

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC510189-1G PC510189-250MG
APOSPC510189-1GEA 1830 DKK
APOSPC510189-1G APOSPC510189-250MG
2-Ethyl-4-fluoro-1-nitrobenzene
2-Ethyl-4-fluoro-1-nitrobenzene
Formel: C₈H₈FNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16658616
CAS-nummer: 1089279-29-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR