2-Bromo-4-(difluoromethoxy)benzonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC510165-1G PC510165-250MG PC510165-5G
APOSPC510165-1GEA 7570 DKK
APOSPC510165-1G APOSPC510165-250MG APOSPC510165-5G
2-Bromo-4-(difluoromethoxy)benzonitrile
2-Bromo-4-(difluoromethoxy)benzonitrile
Formel: C₈H₄BrF₂NO
Molekylvægt: 248.024 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18390874
CAS-nummer: 1261818-72-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR