4-[(Cyclopropylcarbonyl)amino]-3-fluorobenzoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC510053-1G PC510053-5G
APOSPC510053-1GEA 2070 DKK
APOSPC510053-1G APOSPC510053-5G
4-[(Cyclopropylcarbonyl)amino]-3-fluorobenzoic acid
4-[(Cyclopropylcarbonyl)amino]-3-fluorobenzoic acid
Formel: C₁₁H₁₀FNO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18285538
CAS-nummer: 1314406-43-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR