4-Fluorothiophene-2-carboxylic acid 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC50534-100MG PC50534-1G PC50534-250MG
APOSPC50534-100MGEA 1120 DKK
APOSPC50534-100MG APOSPC50534-1G APOSPC50534-250MG
4-Fluorothiophene-2-carboxylic acid 98%
4-Fluorothiophene-2-carboxylic acid
Formel: C₅H₃FO₂S
Opbevaringstemperatur: Køleskab
MDL Number: MFCD10566738
CAS-nummer: 32431-72-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR