5-Bromo-2-fluorophenylisocyanate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
PC503019-1G
PC503019-25G
PC503019-5G
APOSPC503019-1GEA
622
DKK
APOSPC503019-1G
APOSPC503019-25G
APOSPC503019-5G
5-Bromo-2-fluorophenylisocyanate
5-Bromo-2-fluorophenylisocyanate
May form small amount of sediment
Formel:
C₇H₃BrFNO Molekylvægt: 216.007 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09932124 CAS-nummer: 956784-89-7 UN nummer: 2206 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...