4-Bromo-3-fluorophenylisocyanate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC503017-1G PC503017-25G PC503017-5G
APOSPC503017-1GEA 600 DKK
APOSPC503017-1G APOSPC503017-25G APOSPC503017-5G
4-Bromo-3-fluorophenylisocyanate
4-Bromo-3-fluorophenylisocyanate

May form small amount of sediment

Formel: C₇H₃BrFNO
Molekylvægt: 216.007 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD16084774
CAS-nummer: 20059-87-4
UN nummer: 2206
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR