3-Bromo-5-fluorophenylisocyanate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC503012-1G PC503012-25G PC503012-5G
APOSPC503012-1GEA 600 DKK
APOSPC503012-1G APOSPC503012-25G APOSPC503012-5G
3-Bromo-5-fluorophenylisocyanate
3-Bromo-5-fluorophenylisocyanate

May form small amount of sediment

Formel: C₇H₃BrFNO
Molekylvægt: 216.007 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11619406
CAS-nummer: 1094485-14-9
UN nummer: 2206
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR