2-Bromo-6-fluorophenylisocyanate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
PC503003-1G
PC503003-25G
PC503003-5G
APOSPC503003-1GEA
164
DKK
APOSPC503003-1G
APOSPC503003-25G
APOSPC503003-5G
2-Bromo-6-fluorophenylisocyanate
2-Bromo-6-fluorophenylisocyanate
May form small amount of sediment
Formel:
C₇H₃BrFNO Molekylvægt: 216.007 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD22557591 CAS-nummer: 1360960-48-0 UN nummer: 2206 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...