2-Bromo-6-fluorophenylisothiocyanate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC503002-1G PC503002-25G PC503002-5G
APOSPC503002-1GEA 654 DKK
APOSPC503002-1G APOSPC503002-25G APOSPC503002-5G
2-Bromo-6-fluorophenylisothiocyanate
2-Bromo-6-fluorophenylisothiocyanate
Formel: C₇H₃BrFNS
Molekylvægt: 232.073 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22557646
CAS-nummer: 1360888-03-4
UN nummer: 2922
ADR: 8+6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR