2-Bromo-4-fluorophenylisocyanate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

PC502998-1G PC502998-25G PC502998-5G
APOSPC502998-1GEA 164 DKK
APOSPC502998-1G APOSPC502998-25G APOSPC502998-5G
2-Bromo-4-fluorophenylisocyanate
2-Bromo-4-fluorophenylisocyanate

May form small amount of sediment

Formel: C₇H₃BrFNO
Molekylvægt: 216.007 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09929336
CAS-nummer: 1016722-24-9
UN nummer: 2206
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR