2-Fluoro-6-nitrobenzonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC502596-1G PC502596-5G
APOSPC502596-1GEA 933 DKK
APOSPC502596-1G APOSPC502596-5G
2-Fluoro-6-nitrobenzonitrile
2-Fluoro-6-nitrobenzonitrile
Formel: C₇H₃FN₂O₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08063904
CAS-nummer: 143306-27-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR