3-Bromo-5-fluoro-2-methoxyaniline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC502437-10G PC502437-1G PC502437-5G
APOSPC502437-10GEA 4550 DKK
APOSPC502437-10G APOSPC502437-1G APOSPC502437-5G
3-Bromo-5-fluoro-2-methoxyaniline
3-Bromo-5-fluoro-2-methoxyaniline
Formel: C₇H₇BrFNO
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13623651
CAS-nummer: 1096884-89-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR