7-Bromo-6-fluorobenzo[b]thiophene-2-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC501692-1G PC501692-250MG
APOSPC501692-1GEA 4310 DKK
APOSPC501692-1G APOSPC501692-250MG
7-Bromo-6-fluorobenzo[b]thiophene-2-carboxylic acid
7-Bromo-6-fluorobenzo[b]thiophene-2-carboxylic acid
Formel: C₉H₄BrFO₂S
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28967912
CAS-nummer: 1781503-02-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR