2-Fluoro-3-nitro-N-methoxy-N-methylbenzamide
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC501464-1G
PC501464-250MG
PC501464-5G
APOSPC501464-1GEA
5230
DKK
APOSPC501464-1G
APOSPC501464-250MG
APOSPC501464-5G
2-Fluoro-3-nitro-N-methoxy-N-methylbenzamide
2-Fluoro-3-nitro-N-methoxy-N-methylbenzamide
Formel:
C₉H₉FN₂O₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD28397848 CAS-nummer: 1809015-20-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...