4-Bromo-2-fluoro-3-methylaniline 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC501223-1G PC501223-250MG PC501223-5G
APOSPC501223-1GEA 796 DKK
APOSPC501223-1G APOSPC501223-250MG APOSPC501223-5G
4-Bromo-2-fluoro-3-methylaniline 95%
4-Bromo-2-fluoro-3-methylaniline
Formel: C₇H₇BrFN
Molekylvægt: 204.04 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26687842
CAS-nummer: 1540204-53-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR