2-Nitro-3-(trifluoromethyl)benzyl alcohol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC500800-1G PC500800-250MG
APOSPC500800-1GEA 6370 DKK
APOSPC500800-1G APOSPC500800-250MG
2-Nitro-3-(trifluoromethyl)benzyl alcohol
2-Nitro-3-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆F₃NO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16606346
CAS-nummer: 1227511-82-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR