4-Chloro-2-methylphenyl trifluoromethanesulphonate 97%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 5-Chloro-2-{[(trifluoromethyl)sulphonyl]oxy}toluene, 4-Chloro-2-methylphenyl triflate

PC500090-1G PC500090-250MG
APOSPC500090-1GEA 3660 DKK
APOSPC500090-1G APOSPC500090-250MG
4-Chloro-2-methylphenyl trifluoromethanesulphonate 97%
4-Chloro-2-methylphenyl trifluoromethanesulphonate
Formel: C₈H₆ClF₃O₃S
Molekylvægt: 274.645 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD25541852
CAS-nummer: 1446016-75-6
UN nummer: 3265
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR