2-Bromo-4-methoxybenzotrifluoride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-Bromo-4-methoxy-1-(trifluoromethyl)benzene

PC499009-25G PC499009-5G
APOSPC499009-25GEA 2970 DKK
APOSPC499009-25G APOSPC499009-5G
2-Bromo-4-methoxybenzotrifluoride
2-Bromo-4-methoxybenzotrifluoride
Formel: C₈H₆BrF₃O
Molekylvægt: 255.032 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10696314
CAS-nummer: 944901-07-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR