1-Chloro-3,5-dibromo-4-fluorobenzene 98%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC49678-1G PC49678-250MG PC49678-5G
APOSPC49678-1GEA 502 DKK
APOSPC49678-1G APOSPC49678-250MG APOSPC49678-5G
1-Chloro-3,5-dibromo-4-fluorobenzene 98%
1-Chloro-3,5-dibromo-4-fluorobenzene
Formel: C₆H₂Br₂ClF
Molekylvægt: 288.339 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28166159
CAS-nummer: 1805525-99-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR