7-Iodo-5-[(trifluoromethyl)thio]-1,3-benzoxazole
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
7-Iodo-1,3-benzoxazol-5-yl trifluoromethyl sulphide, 7-Iodo-5-[(trifluoromethyl)sulphanyl]-1,3-benzoxazole, 7-Iodo-5-[(trifluoromethyl)thio]benzo[d]oxazole
PC49382-1G
PC49382-250MG
PC49382-5G
APOSPC49382-1GEA
2180
DKK
APOSPC49382-1G
APOSPC49382-250MG
APOSPC49382-5G
7-Iodo-5-[(trifluoromethyl)thio]-1,3-benzoxazole
7-Iodo-5-[(trifluoromethyl)thio]-1,3-benzoxazole
Formel:
C₈H₃F₃INOS Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD26407432 CAS-nummer: 1820736-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...