5-(Pentafluorothio)-1,3-benzoxazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC49305-1G PC49305-250MG PC49305-5G
APOSPC49305-1GEA 2180 DKK
APOSPC49305-1G APOSPC49305-250MG APOSPC49305-5G
5-(Pentafluorothio)-1,3-benzoxazole
5-(Pentafluorothio)-1,3-benzoxazole
Formel: C₇H₄F₅NOS
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22370119
CAS-nummer: 1379812-05-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR