3,5-Difluoro-2-hydroxyphenacyl bromide 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Bromo-1-(3,5-difluoro-2-hydroxyphenyl)ethan-1-one, 2-(Bromoacetyl)-4,6-difluorophenol, 2-Bromo-3',5'-difluoro-2'-hydroxyacetophenone
PC49202-1G
PC49202-5G
APOSPC49202-1GEA
985
DKK
APOSPC49202-1G
APOSPC49202-5G
3,5-Difluoro-2-hydroxyphenacyl bromide 95%
3,5-Difluoro-2-hydroxyphenacyl bromide
Formel:
C₈H₅BrF₂O₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD19053258 CAS-nummer: 1192815-24-9 UN nummer: 3263 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...