1-Chloro-7-fluoroisoquinoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Chloro-7-fluoro-2-azanaphthalene

PC49097-1G PC49097-500MG
APOSPC49097-1GEA 3990 DKK
APOSPC49097-1G APOSPC49097-500MG
1-Chloro-7-fluoroisoquinoline
1-Chloro-7-fluoroisoquinoline
Formel: C₉H₅ClFN
Molekylvægt: 181.594 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16883053
CAS-nummer: 630422-89-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR