2,3-Difluoro-1,4-diiodobenzene
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC49088-1G
PC49088-5G
APOSPC49088-1GEA
665
DKK
APOSPC49088-1G
APOSPC49088-5G
2,3-Difluoro-1,4-diiodobenzene
2,3-Difluoro-1,4-diiodobenzene
Formel:
C₆H₂F₂I₂ Smeltepunkt: 55…60 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD22273735 CAS-nummer: 501433-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...