8-Bromo-6,7-difluoro-1H-3,1-benzoxazine-2,4-dione 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC48999-1G PC48999-250MG
APOSPC48999-1GEA 436 DKK
APOSPC48999-1G APOSPC48999-250MG
8-Bromo-6,7-difluoro-1H-3,1-benzoxazine-2,4-dione 95%
8-Bromo-6,7-difluoro-1H-3,1-benzoxazine-2,4-dione
Formel: C₈H₂BrF₂NO₃
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD31710612
CAS-nummer: 2366994-39-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR