2-(3-(Trifluoromethyl)phenyl)propanoic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC48676-100MG PC48676-1G PC48676-250MG
APOSPC48676-100MGEA 2170 DKK
APOSPC48676-100MG APOSPC48676-1G APOSPC48676-250MG
2-(3-(Trifluoromethyl)phenyl)propanoic acid
2-(3-(Trifluoromethyl)phenyl)propanoic acid
Formel: C₁₀H₉F₃O₂
Molekylvægt: 218.172 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11036891
CAS-nummer: 68718-08-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR