3-Bromo-4,5-difluorophenol 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC48654-1G
PC48654-250MG
APOSPC48654-1GEA
5110
DKK
APOSPC48654-1G
APOSPC48654-250MG
3-Bromo-4,5-difluorophenol 95%
3-Bromo-4,5-difluorophenol
Formel:
C₆H₃BrF₂O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD28556936 CAS-nummer: 1807120-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...