4-Bromo-5-chloro-2-fluoro-N-methoxy-N-methylbenzamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC48481-1G PC48481-250MG
APOSPC48481-1GEA 3270 DKK
APOSPC48481-1G APOSPC48481-250MG
4-Bromo-5-chloro-2-fluoro-N-methoxy-N-methylbenzamide
4-Bromo-5-chloro-2-fluoro-N-methoxy-N-methylbenzamide
Formel: C₉H₈BrClFNO₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28348376
CAS-nummer: 1820740-68-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR