2-Bromo-4-fluorobenzo[b]furan-3(2H)-one

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Bromo-4-fluoro-1-benzofuran-3(2H)-one, 2-Bromo-4-fluorocoumaran-3-one

PC48462-1G PC48462-250MG
APOSPC48462-1GEA 5350 DKK
APOSPC48462-1G APOSPC48462-250MG
2-Bromo-4-fluorobenzo[b]furan-3(2H)-one
2-Bromo-4-fluorobenzo[b]furan-3(2H)-one
Formel: C₈H₄BrFO₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD28096620
CAS-nummer: 1823346-06-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR