1-(2-Chloroethyl)-3-(3-chloro-4-fluoro-phenyl)urea

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC46360-1G PC46360-250MG
APOSPC46360-1GEA 197 DKK
APOSPC46360-1G APOSPC46360-250MG
1-(2-Chloroethyl)-3-(3-chloro-4-fluoro-phenyl)urea
1-(2-Chloroethyl)-3-(3-chloro-4-fluoro-phenyl)urea
Formel: C₉H₉Cl₂FN₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD31372626
CAS-nummer: 848758-99-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR