2,4-Bis(trifluoromethyl)quinoline 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC450561-100MG PC450561-250MG
APOSPC450561-100MGEA 2640 DKK
APOSPC450561-100MG APOSPC450561-250MG
2,4-Bis(trifluoromethyl)quinoline 95%
2,4-Bis(trifluoromethyl)quinoline
Formel: C₁₁H₅F₆N
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28137310
CAS-nummer: 91915-68-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR