1-Bromo-4-fluoro-2-(trichloromethyl)benzene

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC450265-1G PC450265-5G
APOSPC450265-1GEA 1780 DKK
APOSPC450265-1G APOSPC450265-5G
1-Bromo-4-fluoro-2-(trichloromethyl)benzene
1-Bromo-4-fluoro-2-(trichloromethyl)benzene
Formel: C₇H₃BrCl₃F
Molekylvægt: 292.36 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD19442033
CAS-nummer: 1309602-51-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR