Ethyl 3-amino-4,4,4-trifluorobutyrate hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC450211-1G PC450211-5G
APOSPC450211-1GEA 959 DKK
APOSPC450211-1G APOSPC450211-5G
Ethyl 3-amino-4,4,4-trifluorobutyrate hydrochloride
Ethyl 3-amino-4,4,4-trifluorobutyrate hydrochloride
Formel: C₆H₁₀F₃NO₂·ClH
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07784298
CAS-nummer: 146425-31-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR