1,1,1-Tribromotrifluoroacetone

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC450209-1G PC450209-5G
APOSPC450209-1GEA 1320 DKK
APOSPC450209-1G APOSPC450209-5G
1,1,1-Tribromotrifluoroacetone
1,1,1-Tribromotrifluoroacetone
Formel: C₃Br₃F₃O
Kogepunkt: 84,7 °C (743 mm Hg)
Massefylde: 2,7 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18074348
CAS-nummer: 431-91-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR