1-(Perfluorohexyl)docosane 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

PC450134-1G PC450134-250MG
APOSPC450134-1GEA 3060 DKK
APOSPC450134-1G APOSPC450134-250MG
1-(Perfluorohexyl)docosane 95%
1-(Perfluorohexyl)docosane
Formel: C₂₈H₄₅F₁₃
Smeltepunkt: 53…55 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28135666
CAS-nummer: 825651-73-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR