3-(Trifluoromethyl)phenyl fluorosulphonate
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC450050-100MG
PC450050-250MG
APOSPC450050-100MGEA
819
DKK
APOSPC450050-100MG
APOSPC450050-250MG
3-(Trifluoromethyl)phenyl fluorosulphonate
3-(Trifluoromethyl)phenyl fluorosulphonate
Formel:
C₇H₄F₄O₃S Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD24842695 CAS-nummer: 1417220-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...