Ethyl chloro[(4-fluorophenyl)hydrazono]acetate 95%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC430597-1G
PC430597-250MG
APOSPC430597-1GEA
2230
DKK
APOSPC430597-1G
APOSPC430597-250MG
Ethyl chloro[(4-fluorophenyl)hydrazono]acetate 95%
Ethyl chloro[(4-fluorophenyl)hydrazono]acetate
Formel:
C₁₀H₁₀ClFN₂O₂ Smeltepunkt: 109 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06809227 CAS-nummer: 37522-19-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...