2-(3-Fluoropyridin-2-yl)acetonitrile 95%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PC421200-1G PC421200-250MG
APOSPC421200-1GEA 2710 DKK
APOSPC421200-1G APOSPC421200-250MG
2-(3-Fluoropyridin-2-yl)acetonitrile 95%
2-(3-Fluoropyridin-2-yl)acetonitrile
Formel: C₇H₅FN₂
Molekylvægt: 136.126 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09924179
CAS-nummer: 149488-78-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR