5-(Aminomethyl)-2-fluorophenylboronic acid hydrochloride 98%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PC412224-1G
PC412224-250MG
APOSPC412224-1GEA
3450
DKK
APOSPC412224-1G
APOSPC412224-250MG
5-(Aminomethyl)-2-fluorophenylboronic acid hydrochloride 98%
5-(Aminomethyl)-2-fluorophenylboronic acid hydrochloride
Formel:
C₇H₉BFNO₂·ClH Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11053811 CAS-nummer: 1072946-46-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...