5-Fluoro-2-pyridinecarboxamide
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
PC408426-1G
PC408426-25G
PC408426-5G
APOSPC408426-1GEA
183
DKK
APOSPC408426-1G
APOSPC408426-25G
APOSPC408426-5G
5-Fluoro-2-pyridinecarboxamide
5-Fluoro-2-pyridinecarboxamide
Formel:
C₆H₅FN₂O Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11976463 CAS-nummer: 499796-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...